missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse VSIG4 Fc Chimera Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 4674-VS-050

411.22 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18166550

  • 537.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to inhibit anti-CD3-induced proliferation of splenic T lymphocytes. Vogt, L. et al. (2006) J. Clin. Invest. 116:2817. The presence of immobilized Recombinant Mouse VSIG4 Fc Chimera inhibited the anti-CD3 response by =50%. The ED50 for this effect is 1.25-5 μg/mL.
Specifications

Specifications

NP_808457
Lyophilized from a 0.2 μm filtered solution in PBS.
M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 46 kDa (monomer)
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
Reconstitute at 200 μg/mL in sterile PBS.
VSIG4
Bioactivity
11326
VSIG4 Fc Chimera
Mouse myeloma cell line, NS0-derived mouse VSIG4 protein Mouse VSIG4 (His20-Pro187) (N-terminus) Accession # NP_808457 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Disulfide-linked homodimer
Vsig4
Recombinant
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers