missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Aln G 1.0101 Protein

A cDNA sequence encoding the Aln G 1.0101 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11007-1mg

 View more versions of this product

Product Code. 15945848

  • 6300.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Aln G 1.0101 Protein
Research Use Only
Recombinant Protein
Baculovirus
MGVFNYEAET PSVIPAARLF KAFILDGDKL LPKVAPEAVS SVENIEGNGG PGTIKKITFP EGSPFKYVKE RVDEVDRVNF KYSFSVIEGG AVGDALEKVC NEIKIVAAPD GGSILKISNK FHTKGDHEIN AEQIKIEKEK AVGLLKAVES YLLAHSDAYN
Greater than 80.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Aln G 1.0101 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers