missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other LDHB Protein
A cDNA sequence encoding the LDHB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10744-30mg
This item is not returnable.
View return policy
Specifications
LDHB Protein | |
Research Use Only | |
The specific activity was found to be 258 U/mg protein. | |
Other | |
Untagged | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
30 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with 0.1 mg potassium phosphate. |