missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other TBEV NS3 Protein

A cDNA sequence encoding the TBEV NS3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13702-1mg

 View more versions of this product

Product Code. 15954429

  • 1450.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

TBEV NS3 Protein
Research Use Only
Recombinant Protein
E. coli
20mM MES pH 6.5, 8M urea, 200mM NaCl and 0.05% Tween-20.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
Encephalitis protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers