missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant P. vulgaris Chondroitinase ABC Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 6877-GH-020

249.67 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18137810

  • 327.00 € / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to hydrolyze chondroitin sulfate. The specific activity is >15,000 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

P59807
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Brij-35.
M.W. (Observed): 85-100 kDa, reducing conditions; M.W. (theoretical): 113 kDa
20 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
NEWENTRY
Recombinant
Chondroitinase ABC
Enzyme Activity
2827933
Chondroitinase ABC
E. coli-derived p. vulgaris Chondroitinase ABC protein Ala25-Pro1021, with an N-terminal Met and 6-His tag
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers