Learn More
Novex™ Recombinant Protein A-Sepharose™ 4B
Recombinant Protein A-Sepharose™ 4B
Brand: Novex™ 101141
341.05 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
This product is prepared by covalently coupling highly purified Staph. aureus Cowan strain Protein A produced in E. Coli to Sepharose™ 4B.This product is supplied as a 50% suspension consisting of recombinant Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% ).
Applications: Immunoglobulin Affinity Purification, Immunoprecipitation

Specifications
Protein A Sepharose | |
Protein A | |
5 mL |
Affinity | |
2°C to 8°C. Do Not Freeze. This product is supplied as a 50% suspension (5 ml Recombinant Protein A-Sepharose™ 4B + 5 ml PBS) Wet Ice |
|
Suspension |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
QF215070 | Certificate of Analysis | 03/07/2024 | 101141 |
YI375691 | Certificate of Analysis | 06/10/2023 | 101141 |
XL365865 | Certificate of Analysis | 11/01/2023 | 101141 |
XB344698 | Certificate of Analysis | 17/05/2022 | 101141 |
XD349765 | Certificate of Analysis | 17/05/2022 | 101141 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.