missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein G

Recombinant Protein G

163.00 € - 394.00 €

Specifications

Conjugate Unconjugated
Form Powder
Protein Subtype Protein G
Content And Storage 2-8°C
Shipping Condition Wet Ice
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10726713
View Documents
Novex™
101200
1 mg
163.00 €
1mg
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
10100633
View Documents
Novex™
101201
5 mg
394.00 €
5mg
In Stock
to see stock.
Add to basket
 
Description

Description

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. It is supplied as a salt-free lyophilized powder.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • Specifications

    Specifications

    Unconjugated
    Protein G
    Wet Ice
    Powder
    2-8°C
    Videos
    SDS
    Documents

    Documents

    Product Certifications

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.