missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein G - Sepharose™ 4B

Recombinant Protein G - Sepharose™ 4B

Brand:  Novex™ 101241

319.64 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10078792

  • 524.00 € / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This product is prepared by covalently coupling highly purified rec-Protein G to Sepharose™ 4B. It is supplied as a 50% suspension containing rec-Protein G-Sepharose™ 4B + PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin affinity purification and Immunoprecipitation.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Protein G Sepharose
    Affinity
    This product is supplied as a 50% suspension (2 ml Recombinant Protein G-Sepharose™ 4B + 2 ml PBS) Wet Ice
    Suspension
    Sepharose™
    Protein G
    2 mL
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.