Learn More
R&D Systems™ Recombinant Rat Erythropoietin/EPO Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 1306-RE-010
292.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18389287
  • 377.00 € / 10µg
Explore more special offers
Description
Description
The Recombinant Rat Erythropoietin Protein is derived from Sf 21 (stably transfected). The Recombinant Rat Erythropoietin Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
18.5 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Recombinant
24335
Erythropoietin/EPO
Spodoptera frugiperda, Sf 21 (stably transfected)-derived rat Erythropoietin/EPO protein Ala27-Arg192
<0.10 EU per 1μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.