missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Iron-regulated surface determinant protein 1
A DNA sequence encoding the Staphylococcus aureus (strain USA300 / TCH1516) Iron-regulated surface determinant protein B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6999-50ug
This item is not returnable.
View return policy
Specifications
WP_001041586.1 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Iron-regulated surface determinant protein 1 | |
Research Use Only | |
isdB | |
Recombinant Protein | |
E. coli | |
AAEETGGTNT EAQPKTEAVA SPTTTSEKAP ETKPVANAVS VSNKEVEAPT SETKEAKEVK EVKAPKETKE VKPAAKATNN TYPILNQELR EAIKNPAIKD KDHSAPNSRP IDFEMKKKDG TQQFYHYASS VKPARVIFTD SKPEIELGLQ SGQFWRKFEV YEGDKKLPIK LVSYDTVKDY AYIRFSVSNG TKAVKIVSST HFNNKEEKYD YTLMEFAQPI YNSADKFKT | |
Greater than 90% as determined by SDS-PAGE. |