missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant SARS-CoV-2 D614G Spike S1 Subunit His Protein CF
CHO Expressed
Brand: R&D Systems™ 10618-CV-100
516.62 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Recombinant SARS-CoV-2 D614G Spike S1 Subunit His-tag (Catalog # 10618-CV) binds Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ) in a functional ELISA.Specifications
YP_009724390.1 | |
Bioactivity | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
SARS-CoV-2 |
Unconjugated | |
100 μg | |
SARS-CoV-2 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |