missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant SARS-CoV-2 Spike RBD Fc Chimera Protein

CHO Expressed, High ACE-2 Binding

Brand:  R&D Systems™ 10542-CV-100

516.62 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18786706

  • 670.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ).
Specifications

Specifications

YP_009724390.1
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
M.W.-observed: 55-66 kDa, under reducing conditions, M.W.-theoretical: 52 kDa
Chinese Hamster Ovary cell line, CHO-derived sars-cov-2 Spike RBD protein SARS-CoV2 Spike RBD (Arg319-Phe541) (N-terminus) Accession # YP_009724390.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
<0.10 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Bioactivity
43740568
100 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein
Reconstitute at 500 μg/mL in PBS.
SARS-CoV-2 Spike RBD
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers