Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike S1 HR1/HR2 His Protein, CF
Carrier-Free Recombinant Protein
Brand:  R&D Systems™ 10687-CV-100
483.79 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18166280
  • 614.00 € / 100µg
Explore more special offers
Description
Description
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 His-tag (Catalog # 933-ZN ).
Specifications
Specifications
YP_009724390.1
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
100 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
SARS-CoV-2
Reconstitute at 500 μg/mL in PBS.
SARS-CoV-2 Spike S1 Protein
Bioactivity
M.W. (Observed): 125-145 kDa, under reducing conditions, M.W. (Theoretical): 92 kDa
Chinese Hamster Ovary cell line, CHO-derived sars-cov-2 Spike S1 Subunit protein SARS-CoV-2 Spike S1 (Val16-Pro681) (N-terminus) Accession # YP_009724390.1 HR1/HR2 HHHHHH (C-terminus)
<0.10 EU per 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.