Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike S1 Subunit Biotin Protein
His-tag. Biotinylated via Amines
Brand:  R&D Systems™ BT10569-050
596.10 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 30000916
  • 779.00 € / 50µg
Explore more special offers
Description
Description
Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN).
Specifications
Specifications
YP_009724390.1
Bioactivity
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
SARS-CoV-2
Biotin
50 μg
SARS-CoV-2
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.