missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant SARS-CoV-2 Spike S2 (GCN4-IZ) His Protein

GCN4-IZ trimerization domain,

Brand:  R&D Systems™ 10590-CV-100

516.62 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 30000969

  • 656.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Bioassay data are not available.
Specifications

Specifications

YP_009724390.1
Bioactivity
100 μg
COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Unconjugated
43740568
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
SARS-CoV-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers