missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Alexa Fluor 488, Clone: CTK0101 and CTK0102, Proteintech ChromoTek
Alpaca Monoclonal Antibody
91.00 € - 318.00 €
Specifications
Antigen | Recombinant VHH |
---|---|
Clone | CTK0101 and CTK0102 |
Applications | Immunofluorescence, Western Blot, Flow Cytometry |
Host Species | Alpaca |
Target Species | Human, Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17353493
|
Chromotek
SRBAF488-1-10 |
10 μL |
91.00 €
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17363493
|
Chromotek
SRBAF488-1-100 |
100 μL |
318.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nano-Secondary™ alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Specifications
Recombinant VHH | |
Immunofluorescence, Western Blot, Flow Cytometry | |
Human, Rabbit |
CTK0101 and CTK0102 | |
Alpaca |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title