missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 488, Clone: CTK0103 and CTK0104, Proteintech ChromoTek
Alpaca Monoclonal Antibody
90.00 € - 329.68 €
Specifications
Antigen | Recombinant VHH |
---|---|
Clone | CTK0103 and CTK0104 |
Applications | Immunofluorescence, Western Blot, Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17393463
|
Chromotek
sms1AF488-1-10 |
10 μL |
90.00 €
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17303473
|
Chromotek
SMS1AF488-1-100 |
100 μL |
329.68 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nano-Secondary™ alpaca anti-mouse IgG1, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG1 with high affinity & specificity.
Specifications
Recombinant VHH | |
Immunofluorescence, Western Blot, Flow Cytometry | |
Alexa Fluor 488 | |
Mouse | |
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
CTK0103 and CTK0104 | |
Monoclonal | |
Alpaca | |
Secondary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 488, Clone: CTK0103 and CTK0104, Proteintech ChromoTek