Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 488, Clone: CTK0105 and CTK0106, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand:  Chromotek SMS2BAF488-1-10
 View more versions of this product
Product Code. 17353473
  • 91.00 € / 10µL
Explore more special offers
Description
Description
Nano-Secondary™ alpaca anti-mouse IgG2b, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG2b with high affinity & specificity.
Specifications
Specifications
Recombinant VHH
CTK0105 and CTK0106
10 μL
Immunofluorescence, Western Blot
Alpaca