Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 488, Clone: CTK0105 and CTK0106, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand:  Chromotek sms2bAF488-1-100
 View more versions of this product
Product Code. 17363473
  • 318.00 € / 100µL
Explore more special offers
Description
Description
Nano-Secondary™ alpaca anti-mouse IgG2b, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG2b with high affinity & specificity.
Specifications
Specifications
Recombinant VHH
CTK0105 and CTK0106
100 μL
Immunofluorescence, Western Blot
Alpaca