missing translation for 'onlineSavingsMsg'
Learn More

Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0103 and CTK0104, Proteintech ChromoTek

Alpaca Monoclonal Antibody

Brand:  Chromotek sms1AF647-1-100

 View more versions of this product

Product Code. 17343473

  • 319.03 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nano-Secondary™ alpaca anti-mouse IgG1, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG1 with high affinity & specificity.

Specifications

Specifications

Recombinant VHH
Monoclonal
Alexa Fluor 647
100 μL
Mouse
Immunofluorescence, Western Blot, Flow Cytometry
CTK0103 and CTK0104
Alpaca
Secondary
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.