missing translation for 'onlineSavingsMsg'
Learn More

Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0105 and CTK0106, Proteintech ChromoTek

Alpaca Monoclonal Antibody

Brand:  Chromotek SMS2BAF647-1-100

 View more versions of this product

Product Code. 17303483

  • 318.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Nano-Secondary™ alpaca anti-mouse IgG2b, Fc-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of Nanobodies that bind to mouse IgG2b with high affinity & specificity.

Specifications

Specifications

Recombinant VHH
Monoclonal
Alexa Fluor 647
100 μL
Mouse
Immunofluorescence, Western Blot
CTK0105 and CTK0106
Alpaca
Secondary
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers