missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant VHH Alpaca, anti-Mouse, Alexa Fluor 647, Clone: CTK0107, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand: Chromotek sms3AF647-1-10
This item is not returnable.
View return policy
Description
Nano-Secondary™ alpaca anti-mouse IgG3, Fab-specific recombinant VHH is an anti-mouse IgG subclass specific antibody. This secondary antibody product consists of a Nanobody that binds to mouse IgG3 with high affinity & specificity.
Specifications
Recombinant VHH | |
Monoclonal | |
Alexa Fluor 647 | |
10 μL | |
Mouse |
Immunofluorescence, Western Blot | |
CTK0107 | |
Alpaca | |
Secondary | |
Store at -20°C short term or -80°C long term. Aliquot upon delivery. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction