Learn More
Recombinant VHH Alpaca, anti-Rabbit, Unconjugated, Clone: CTK0102, Proteintech ChromoTek
Alpaca Monoclonal Antibody
Brand: Chromotek srbGCys2-2-500
This item is not returnable.
View return policy
Description
500 μg Nano-Secondary™ for rabbit IgG: alpaca monoclonal Nanobody, Fab-specific VHH, for 2x Cys conjugation, clone [CTK0102].
Specifications
Recombinant VHH | |
CTK0102 | |
500 μg |
Immunofluorescence, Western Blot, Flow Cytometry | |
Alpaca | |
Rabbit |