missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RecoverAll™ Multi-Sample RNA/DNA Isolation Workflow

Product Code. p-7076203
Click to view available options
Quantity:
120 Preps
200 Preps
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15430994

Brand: Invitrogen™ A26135

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The RecoverAll™ Multi-Sample RNA/DNA Workflow is a complete workflow package for sequential extraction of RNA and DNA from the same formaldehyde- or paraformaldehyde-fixed paraffin-embedded (FFPE) tissue sample.

The RecoverAll™ Multi-Sample RNA/DNA Workflow is a complete workflow package for sequential extraction of RNA and DNA from the same formaldehyde- or paraformaldehyde-fixed paraffin-embedded (FFPE) tissue sample. The recovered RNA and DNA are ready for use in downstream applications such as quantitative real-time RT-PCR and targeted sequencing using the Oncomine™ Cancer Research Panel. This workflow package includes RecoverAll™ Total Nucleic Acid Isolation kits, elution tubes, and nuclease-free water.

Features include:
• Recovers RNA and DNA in separate eluates from the same FFPE section
• Relies on proven Ambion™ RecoverAll technology
• Processes four 20-μm sections per reaction

Special workflow provides more information from less sample input
FFPE samples are precious, which often requires the choosing of one extraction method over another to save sample. With the RecoverAll Multi-Sample RNA/DNA Workflow, recovering RNA and DNA from the same FFPE tissue sample in a single workflow eliminates the need to choose which nucleic acid to recover from a particular FFPE section.

First, FFPE slide-mounted or tissue blocks sections are deparaffinized and digested with protease. The samples are then loaded on a filter cartridge; DNA remains bound to the filter cartridge while RNA flows through. To recover RNA, the flow-through is applied to a new filter cartridge, treated with DNase, then eluted from the filter. To recover DNA, the DNA bound to the filter cartridge is treated with RNase A, then eluted from the filter.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 5 RecoverAll Total Nucleic Acid Isolation kits (shipped in two parts: one at room temperature and one at -20°C; store as indicated on box)
• 100 elution tubes
• 100 mL nuclease-free water
Sample Type FFPE and Fixed Samples, Fixed Samples
High-throughput Compatibility Not High-throughput Compatible
For Use With (Application) PCR, Northern Blotting, RT-PCR (Endpoint), microRNA Analysis, Real-Time PCR, Sequencing
No. of Reactions 200 Preps
Product Line Ambion, RecoverAll
Quantity 200 Preps
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Target Total RNA, miRNA, Total Nucleic Acids, mRNA, Genomic DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.