Learn More
Gilac™ Rectangular HACCP Container with Lid
Rectangular HACCP Container with Lid is a food container for large storage volumes; lid can support the weight of heavy loads.
38.65 € - 99.95 €
Specifications
Type Storage Tote
Material HDPE
Shape Rectangular
View More Specs
Products 9
Product Code Brand Color Capacity (Metric) Price Quantity & Availability  
Product Code Brand Color Capacity (Metric) Price Quantity & Availability  
17978421
View Documents
Gilac™
G119509
Red 12 L
38.65 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17968561
missing translation for 'viewDocuments'
Gilac™
G119518
Yellow 12 L
38.65 €
Each
In Stock
to see stock.
Edge
Add to basket
17908621
missing translation for 'viewDocuments'
Gilac™
G119612
White 15 L
46.40 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17988811
missing translation for 'viewDocuments'
Gilac™
G119611
Green 15 L
46.40 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17998561
missing translation for 'viewDocuments'
Gilac™
G119609
Red 15 L
46.40 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17938621
missing translation for 'viewDocuments'
Gilac™
G119618
Yellow 15 L
46.40 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17958751
missing translation for 'viewDocuments'
Gilac™
G119809
Red 35 L
75.70 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17928471
missing translation for 'viewDocuments'
Gilac™
G119818
Yellow 35 L
75.70 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
17958861
missing translation for 'viewDocuments'
Gilac™
G119911
Green 55 L
99.95 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
Description
Description
  • HACCP rectangular food container.
  • For large storage volumes, lid can support the weight of heavy loads.
  • Multipurpose container.
  • Rewritable identification cartridge for inventory tracking.
  • No gaps nor overlaps.
  • Machine washable.
  • Recessed handles for better handling.
  • Stackable without lid.
  • Compatible with all GILAC 4 and 6 wheel dollies.
Specifications
Specifications
Storage Tote
HDPE
Rectangular
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Gilac™ Rectangular HACCP Container with Lid