missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Redox Solution, 600mV Pt/Calomel, 640mV Pt/Ag/AgCl, SI Analytics™
Click to view available options
Quantity:
60 Ampules
Description
- GHS07: Skin Irrit. 2 H315 Causes skin irritation, Eye Irrit. 2 H319 Causes serious eye irritation
- Precautionary statements: P280 Wear protective gloves / eye protection / face protection. P264 Wash thoroughly after handling. P305+P351+P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P321 Specific treatment (see on this label). P332+P313 If skin irritation occurs: Get medical advice/attention. P337+P313 If eye irritation persists: Get medical advice/attention. P302+P352 IF ON SKIN: Wash with plenty of water.
- Ingredients with limit values that require monitoring at the workplace: 7664-93-9 sulphuric acid
Specifications
Specifications
Boiling Point | 102°C |
Color | Light Yellow |
Density | 1.15g/cm3 |
pH | -0.2 |
Quantity | 60 Ampules |
Solution Type | Redox |
Odor | Odorless |
Vapor Pressure | 23hPa |
Solubility Information | Fully miscible in water |
Safety and Handling
Product Identifier
- Redox test solution (+600 mV+640 mV)
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362-Take off contaminated clothing.
- P364-And wash it before reuse.
- MIXTURE LIST-Contains : sulphuric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'