Learn More
Lenz Laborglasintrumente Reduction Adapters with Ground Joints
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01504560
This item is not returnable.
View return policy
Description
- Acc. to DIN 12257 and in addition to DIN standard
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifications
NS 60/46 | |
Borosilicate Glass (3.3) | |
Socket NS 45/40 |
Product Content Correction
Product Title
Lenz Laborglasintrumente Reduction Adapters with Ground Joints > Socket NS 45/40
Spot an opportunity for improvement?Share a Content Correction