Learn More
Lenz Laborglasintrumente Reduction Adapters with Ground Joints
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01504534
 View more versions of this product
Product Code. 10385451
  • 56.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12257 and in addition to DIN standard
  • Conical ground sockets and cones
  • Made of DURAN™ tubing
Specifications
Specifications
NS 34/35
Borosilicate Glass (3.3)
Socket NS 45/40
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Reduction Adapters with Ground Joints > Socket NS 45/40