Learn More
asecos™ Refill Kit for Trolleys Sorbents
Product Code. 17806456 Shop All asecos Products
Click to view available options
Type:
Chemical Spill Control Absorbent Storage Cart Refill Kit
Oil Spill Control Absorbent Storage Cart Refill Kit
Universal All Purpose Absorbent Storage Cart Refill Kit
Unit Size:
Each
Product Code. 17806456
Brand: asecos™ 14415
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Refill Kit for Trolleys Sorbents is a practical Refill Set for trolley spill kits.
  • content: 6 cushions, 3 socks, 20 mats, 3 pair protective gloves, 5 plastic bags
Specifications
Material Heavy-duty Cloth
Type Universal All Purpose Absorbent Storage Cart Refill Kit
Weight (English) 13.23 lb.
Weight (Metric) 6 kg