Learn More
Labcon™ Refillable Box for 192 Pipette Stacking Racks
Refillable box for storing racked pipette tips.
Brand:  Labcon™ 1215-120-000-9
Product Code. 17557876
  • 12.56 € / Pack of 5
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Refillable rack with hinged clear lid
  • Accepts two Labcon™ 8 x 24 pipette floating racks
  • Freezable to -80°C
  • Made in USA
Specifications
Specifications
ISO 9001:2015, FDA CFR21 GMP
Freezer Storage Box
Autoclavable
Polypropylene
Yes
Floating Racks
5/Pk.
Hinged Box without Racks
8 x 24
White
Rectanglular
Non-sterile
50/Cs.
5 Boxes/Pk., 10 Pk./Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labcon™ Refillable Box for 192 Pipette Stacking Racks > Refillable Box for 192 Pipette Stack Racks; Array: 8 x 24; Certifications/Compliance: ISO 9001:2015, FDA CFR21 GMP; For Use With (Equipment): Floating Racks; Color: White; Material: Polypropylene