Learn More
Brady™ Reflective Labels (B-584)
Labeling in low light conditions. Brady Reflective Labels (B-584) are for use with BMP71 printer.
Brand:  Brady™ 104304
 View more versions of this product
Product Code. 11899524
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Reflective tape for use in dark area
  • Available in yellow, orange and silver
Specifications
Specifications
Class 1 (Fed. Spec. L-S-300)
Acrylic, Permanent
Gloss
Reflective Tape
BMP™71 Printer
1 Ea.
0.0055 in.
75 ft.
2 in.
Suggested ribbon series: R6000
Yellow
Reflective Tape (B-584)
Pedestal Identification
1
-40°C to +70°C
0.14 mm
22.8 m
5.08 cm