missing translation for 'onlineSavingsMsg'
Learn More

Reformulated Organohalide Pesticides Mix, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep 505-A2


Product Code. 16349787

  • 74.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
This item is not returnable. View return policy

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Atrazine1912-24-91250μg/mL
Simazine122-34-91250μg/mL
Alachlor15972-60-850μg/mL
Methoxychlor72-43-525μg/mL
Aldrin309-00-25μg/mL
alpha-Chlordane5103-71-95μg/mL
cis-Nonachlor5103-73-15μg/mL
Dieldrin60-57-15μg/mL
Endrin72-20-85μg/mL
gamma-BHC58-89-95μg/mL
gamma-Chlordane5103-74-25μg/mL
Heptachlor76-44-85μg/mL
Heptachlor epoxide (Isomer B)1024-57-35μg/mL
Hexachlorobenzene118-74-15μg/mL
Hexachlorocyclopentadiene77-47-45μg/mL
trans-Nonachlor39765-80-55μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Reformulated Organohalide Pesticides Mix
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For US EPA Method 505
USEPA Method 505
1 mL
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Acetone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Reformulated Organohalide Pesticides Mix

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : acetone, atrazine, simazine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Product intended for laboratory use only