Learn More
Bellingham Stanley™ Refractometer Calibration Oil
Bellingham Stanley™ Refractometer Calibration Oils cover the range 1.47 -1.56 RI and are are ideal for calibrating refractometers that have temperature control in both food and industrial applications.
Brand:  Bellingham Stanley™ 90-531
Product Code. 16299421
  • 177.00 € / Pack of 5
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For refractometer calibration
  • A mix of organic compounds that cover the range 1.47 -1.56 RI at the top end of the Brix scale
  • Sold as a pack of five 5 mL glass bottles
  • Includes plastic pipette
  • 12-months shelf life
  • Traceable to NIST
  • Supplied with UKAS certificate of calibration
  • Value to five decimal places
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calibration Oil
Refractometer
Dropping pipette
1.53625
±0.000074 RI
5 x 5 mL