Learn More
Techne™ Refrigerated Bath
Handles wide range of temperatures
4030.00 € - 4715.00 €
Specifications
Features Circulating bath with integrated refrigeration unit, Temperature range from -35°C to +100°C, Robust stainless-steel construction, Three models of refrigerated bath available: 7, 12, 22 liter,3 year warranty.
Warranty 3 Years
Voltage 230 V
Frequency 50/60 Hz
View More Specs
Products 2
Product Code Brand Model Capacity (Metric) Price Quantity & Availability  
Product Code Brand Model Capacity (Metric) Price Quantity & Availability  
11553769
View Documents
Techne™
12212-04
RB-5A 7 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11543769
View Documents
Techne™
12212-02
RB-12A 12 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Built-in refrigeration for efficient cooling
  • Robust stainless steel construction makes for easy cleaning
Specifications
Specifications
Circulating bath with integrated refrigeration unit, Temperature range from -35°C to +100°C, Robust stainless-steel construction, Three models of refrigerated bath available: 7, 12, 22 liter,3 year warranty.
3 Years
230 V
50/60 Hz