Learn More
Lonza™ REGM™ Renal Epithelial Cell Growth Medium SingleQuots™ Kit
Supplements and growth factors kit for use with renal epithelial cell basal medium.
Brand: Lonza™ CC-4127
This item is not returnable.
View return policy
Description
- Separately sold SingleQuot™ kit for supplementing basal medium.
- Part of the full REGM™ Renal Epithelial Cell Growth Medium BulletKit™ (catalog number CC-3190), which consists of basal medium and supplements packaged together.
- Basal medium and SingleQuot™ kit formulated to maintain and expand most renal epithelial cells.
- Includes the following:
- 1 x Green Cap Vial with hEGF, 0.50 mL
- 1 x Lilac Cap Vial with Insulin, 0.50 mL
- 1 x Natural Cap Vial with Hydrocortisone, 0.50 mL
- 1 x Red Cap Vial with GA-1000, 0.50 mL
- 1 x Bottle with FBS, 2.50 mL
- 1 x Natural Cap Vial with Transferrin, 0.50 mL
- 1 x Amber Vial with Triiodothyronine, 0.50 mL
- 1 x Amber Vial with Epinephrine, 0.50 mL
Specifications
hEGF, Insulin, Hydrocortisone, GA-1000, FBS, Transferrin, Triiodthyronine, Epinephrine | |
Epithelial Cells | |
Supplements and growth factors | |
hEGF: 0.5 mL, Insulin: 0.5 mL, Hydrocortisone: 0.5 mL, GA-1000: 0.5 mL, FBS: 2.5 mL, Transferrin: 0.5 mL, Triiodothyronine: 0.5 mL, Epinephrine: 0.5 mL |