Learn More
Reichert™ Auxiliary Magnifying Lens
For strain-free visual identification of colonies
Brand:  Reichert™ 13333100
Product Code. 10546285
  • 453.00 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
For strain-free visual identification of colonies, doubles the total magnification from 1.5X to 3.0X
Colony counter lens
1.5x
Lens, Auxiliary Magnifying