Learn More
Reichert™ Reichert™ Rhino Ultra-Durable Handheld Refractometers
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
Brand:  Reichert™ 137531L0
 View more versions of this product
Product Code. 10309714
  • 272.00 € / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
  • Automatic Temperature Compensation eliminates the need for temperature correction charts
  • Optics and instrument scales are permanently sealed inside the refractometer body
  • Magnified scales have easy-to-read markings and divisions for true readings and accurate results
  • Models are offered with Brix scales of 0-30, 0-50 and 50-90%
Order Info
Rhino Model for Industrial Fluid Testing is also available; see Cat. No. RE-10440.
Specifications
Specifications
Handheld Ultra-durable Refractometer
1.333 to 1.42
0.25%
IIP67
0 to 50 Brix
±0.25
Food and beverage, chemical, pharmaceutical and industrial cutting fluids