Learn More
Reichert™ Reichert™ Rhino Ultra-Durable Handheld Refractometers
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
Brand:  Reichert™ 137530L0
 View more versions of this product
Product Code. 10546475
  • 272.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed specifically for rugged factory and field use—fully IIP67 water rresistant, dustproof, and impact resistant
  • Automatic Temperature Compensation eliminates the need for temperature correction charts
  • Optics and instrument scales are permanently sealed inside the refractometer body
  • Magnified scales have easy-to-read markings and divisions for true readings and accurate results
  • Models are offered with Brix scales of 0-30, 0-50 and 50-90%
Order Info
Rhino Model for Industrial Fluid Testing is also available; see Cat. No. RE-10440.
Specifications
Specifications
Handheld Ultra-durable Refractometer
1.333 to 1.3811
0.1%
Food and beverage, chemical, pharmaceutical and industrial cutting fluids
9.7 x 1.5 x 1.5 in. (246.3 x 38.1 x 38.1 mm)
38.1 mm
246.3 mm
38.1 mm
0 to 30 Brix
±0.2
0.2 and 0.5
IP67
1.5 in.
9.7 in.
1.5 in.