Learn More
Memmert™ Reinforced Stainless Steel Shelf
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP750eco
IPP1060eco
IPP750eco
Unit Size:
Each
Description
- With guide bars and fixing screws
- Requires option K1
- Consider max. loading of chamber; max. loading 60 kg
Specifications
Specifications
| For Use With (Equipment) | HPP750eco |
| Material | Stainless Steel |