Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Product Code. p-4528205
Click to view available options
Description:
E. coli Polyvalent 2
E. coli Polyvalent 3
E. coli Polyvalent 4
Unit Size:
Each
Product Code. 12647539
Brand: Thermo Scientific™ R30955101
This item requires a price update prior to being ordered.
Request a Price for this item here.
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
TRUSTED_SUSTAINABILITY
Specifications
Description E. coli Polyvalent 4
Detectable Analytes E. coli
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Specificity types O44, O112, O124, O142