Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Brand: Thermo Scientific™ R30166301
This item is not returnable.
View return policy
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
Agglutinating Sera, Haemophilus influenzae Type D | |
H. influenzae | |
Agglutinating Sera |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Haemophilus influenzae Agglutinating Sera
Version: FEB.2016
File size: 530.5 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6127644 | Certificate of Analysis | 06/11/2024 | R30166301 |
3800013 | Certificate of Analysis | 06/04/2024 | R30166301 |
3753271 | Certificate of Analysis | 07/02/2024 | R30166301 |
3671601 | Certificate of Analysis | 21/10/2023 | R30166301 |
3644900 | Certificate of Analysis | 26/05/2023 | R30166301 |
Product Content Correction
Product Title
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae > Type D
Spot an opportunity for improvement?Share a Content Correction