Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Brand: Thermo Scientific™ R30166401
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
Agglutinating Sera, Haemophilus influenzae Type E | |
H. influenzae | |
Agglutinating Sera |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6217481 | Certificate of Analysis | 27/03/2025 | R30166401 |
6002008 | Certificate of Analysis | 19/10/2024 | R30166401 |
3800014 | Certificate of Analysis | 04/06/2024 | R30166401 |
3790181 | Certificate of Analysis | 07/02/2024 | R30166401 |
3702498 | Certificate of Analysis | 24/10/2023 | R30166401 |