missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H

Product Code. 12697429
Click to view available options
Description:
Polyvalent, Phases 1 and 2
Rapid Diagnostic 1 (b, d, E, r)
Rapid Diagnostic 2 (b, E, k, L)
Rapid Diagnostic 3 (d, E, G, k)
Salmonella H (1, 2)
Salmonella H (1, 5)
Salmonella H (1, 6)
Salmonella H (1, 7)
Salmonella H (L) Polyvalent
Salmonella H (a)
Salmonella H (b)
Salmonella H (c )
Salmonella H (d)
Salmonella H (eh)
Salmonella H (enx)
Salmonella H (f, g)
Salmonella H (gm)
Salmonella H (r )
Salmonella H (y)
Salmonella H (z27)
Unit Size:
2mL
Each
This item is not returnable. View return policy

Product Code. 12697429

Brand: Thermo Scientific™ R30163601

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.

Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.

  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Salmonella H (1, 7)
CE Marker Yes
Detectable Analytes Salmonella H Antigen
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
Target Organism Class Salmonella
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.