Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Brand:  Thermo Scientific™ R30161801
 View more versions of this product
Product Code. 12719300
  • 163.00 € / Each
Explore more special offers
Description
Description
Remel Agglutinating Sera, Salmonella H is used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Positive
Salmonella H Antigen
Polyvalent
2 mL
Salmonella H (L) Polyvalent, for lv, lw, lz13, lz28, lz38, lz40
Agglutinating Sera
2 mL/Vial
Salmonella H