Learn More
Thermo Scientific™ Remel™ Gram Crystal Violet
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
59.25 € - 156.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11982952
|
Thermo Scientific™
R40052 |
Each |
59.25 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12663658
|
Thermo Scientific™
R40053 |
5/Pk. | N/A | N/A | N/A | |||||
11914092
|
Thermo Scientific™
R40073 |
Each |
156.00 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
- For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
- Selected Gram stains are CE marked
Specifications
Crystal Violet | |
CE Marked |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Remel™ Gram Crystal Violet