Learn More
Thermo Scientific™ Remel™ Gram Iodine
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
56.55 € - 239.00 €
Specifications
Type Gram Iodine
Certifications/Compliance CE Marked
View More Specs
Products 3
Product Code Brand Type Quantity Certifications/Compliance Price Quantity & Availability  
Product Code Brand Type Quantity Certifications/Compliance Price Quantity & Availability  
11983322
View Documents
Thermo Scientific™
R40077
Gram Iodine Each CE Marked This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11933972
View Documents
Thermo Scientific™
R40056
Gram Iodine Each CE Marked
56.55 €
250mL
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
12687589
View Documents
Thermo Scientific™
R40057
Gram Iodine 5/Pk. CE Marked This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
Specifications
Specifications
Gram Iodine
CE Marked