missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 2
Remel PathoDx and Extraction Reagent 2 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
28.20 € - 30.05 €
Specifications
Content And Storage | 2°C to 8°C |
---|---|
CE Marker | Yes |
Format | Vial |
Quantity | Each |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10402123
|
Thermo Scientific™
R62055 |
PathoDx Extraction Reagent 2, 7mL/Vial | Each |
28.20 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12771770
|
Thermo Scientific™
R62057 |
PathoDx Extraction Reagent 2, 14mL/Vial | Each |
30.05 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Remel PathoDx and Extraction Reagent 2 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
Specifications
2°C to 8°C | |
Yes | |
Vial | |
Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title