Learn More
Thermo Scientific™ PathoDx™ Respiratory Virus Panel
PathoDx Respiratory Virus Panel (RVP) kit is a direct immunofluorescence test for the qualitative detection of 7 common respiratory viruses.
Brand:  Thermo Scientific™ R62400
Product Code. 11424129
Description
Show More
Specifications
Specifications
Respiratory Virus; 200 Tests/Kit
Respiratory Syncytial Virus (RSV)
Panel
200 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3380346 Certificate TypeCertificate of Analysis Date01/04/2022 Catalog NumberR62400
Lot Number3376349 Certificate TypeCertificate of Analysis Date23/12/2021 Catalog NumberR62400
Lot Number3245345 Certificate TypeCertificate of Analysis Date01/06/2021 Catalog NumberR62400
Lot Number2969050 Certificate TypeCertificate of Analysis Date17/06/2020 Catalog NumberR62400
Lot Number2919300 Certificate TypeCertificate of Analysis Date11/03/2020 Catalog NumberR62400
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ PathoDx™ Respiratory Virus Panel > Respiratory Virus; 200 tests/kit