missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, pH 5.6

Remel Sabouraud Dextrose Agar, pH 5.6 is used for the isolation of fungi.

Brand:  Thermo Scientific™ R454462

 View more versions of this product

Product Code. 12677619

  • 122.00 € / 500g
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Remel Sabouraud Dextrose Agar, pH 5.6 is used for the isolation of fungi.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: USP
Powder
500 g
Sabouraud Dextrose Agar, pH 5.6
Agar
For 7.7L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Laboratory Use Only