missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ XLD Agar

Remel XLD Agar is used for the isolation and differentiation of enteric pathogens.

Brand:  Thermo Scientific™ R459902

Product Code. 12968244

  • 356.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Remel XLD Agar is used for the isolation and differentiation of enteric pathogens.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Industrial Reference: USP
Powder
500 g
XLD Agar
Agar
For 9.0L medium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number209964 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberR459902
Lot Number127943 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberR459902
Lot Number997137 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR459902
Lot Number758520 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR459902
Lot Number621129 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberR459902
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Remel™ XLD Agar > XLD Agar; 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only